Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, single neck, 150mL, 24/40
SDP
Catalog No. 50973498
Encompass_Preferred
Flask, round bottom, heavy wall, single neck, 150mL, 24/40
Catalog No. 50-973-498 Supplier Ace Glass, Inc. Supplier No. 6887233
Flask, round bottom, heavy wall, single neck, 150mL, 24/40