Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, 1L, 24/40 center, 24/40side
SDP
Supplier:  Ace Glass, Inc. 6928232
Encompass_Preferred
1L heavy wall round bottom flask with one center neck and one vertical side neck, both with 24/40 standard taper joints.
Catalog No. 50-978-938