Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, two neck, 1L, 24/40 center, 24/40 side
SDP
Catalog No. 50973586
Encompass_Preferred
1L two neck, round bottom heavy wall flask with standard joints. Center neck and angled side neck both have 24/40 joints.
Catalog No. 50-973-586 Supplier Ace Glass, Inc. Supplier No. 6927232
1L two neck, round bottom heavy wall flask with standard joints. Center neck and angled side neck both have 24/40 joints.