Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, 1L, 24/40 center, stopcock septum port side arm
SDP
Catalog No. 50973609
Encompass_Preferred
1L heavy wall round bottom flask with a single 24/40 center neck, and a stopcock septum port side arm, for use with air-sensitive material. Flask features a PTFE 2mm bore stopcock. Supplied with a red rubber septum.
Catalog No. 50-973-609 Supplier Ace Glass, Inc. Supplier No. 6934229
1L heavy wall round bottom flask with a single 24/40 center neck, and a stopcock septum port side arm, for use with air-sensitive material. Flask features a PTFE 2mm bore stopcock. Supplied with a red rubber septum.