Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, three neck, 1L, 29/42 center, 29/42 sides
SDP
Catalog No. 50973686
Encompass_Preferred
1L heavy wall, round bottom flask with three vertical necks. Center neck and side necks have 29/42 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.
Catalog No. 50-973-686 Supplier Ace Glass, Inc. Supplier No. 6944228
1L heavy wall, round bottom flask with three vertical necks. Center neck and side necks have 29/42 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.