Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, 2L, 24/40 center, 24/40side
SDP
Catalog No. 50973596
Encompass_Preferred
2L heavy wall round bottom flask with one center neck and one vertical side neck, both with 24/40 standard taper joints.
Catalog No. 50-973-596 Supplier Ace Glass, Inc. Supplier No. 6928244
2L heavy wall round bottom flask with one center neck and one vertical side neck, both with 24/40 standard taper joints.