Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, three neck, 2L, 24/40 center, 24/40 sides
SDP
Catalog No. 50973690
Encompass_Preferred
2L heavy wall, round bottom flask with three vertical necks. Center neck and side necks have 24/40 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.
Catalog No. 50-973-690 Supplier Ace Glass, Inc. Supplier No. 6944240
2L heavy wall, round bottom flask with three vertical necks. Center neck and side necks have 24/40 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.