Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, four neck, 2L, 29/42 center, (2)24/40, (1) #7 sides
SDP
Catalog No. 50973875
Encompass_Preferred
2L heavy wall, round bottom flask with four necks, arranged three in-line, and one #7 Ace-Thred neck. Center neck has 29/40 center joint, and angled side necks have 24/40 joints. Supplied with bushing and FETFE o-ring.
Catalog No. 50-973-875 Supplier Ace Glass, Inc. Supplier No. 6954283
2L heavy wall, round bottom flask with four necks, arranged three in-line, and one #7 Ace-Thred neck. Center neck has 29/40 center joint, and angled side necks have 24/40 joints. Supplied with bushing and FETFE o-ring.