Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, 2L, 29/42 center, 24/40side
SDP
Catalog No. 50973597
Encompass_Preferred
2L heavy wall round bottom flask with one center neck with a 29/42 standard taper joint and one vertical side neck with a 24/40 standard taper joint.
Catalog No. 50-973-597 Supplier Ace Glass, Inc. Supplier No. 6928246
2L heavy wall round bottom flask with one center neck with a 29/42 standard taper joint and one vertical side neck with a 24/40 standard taper joint.