Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, three neck, 2L, 34/45 center, 24/40 sides
SDP
Catalog No. 50973693
Encompass_Preferred
2L heavy wall, round bottom flask with three vertical necks. Center neck has 34/45 joint, and side necks have 24/40 joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.
Catalog No. 50-973-693 Supplier Ace Glass, Inc. Supplier No. 6944246
2L heavy wall, round bottom flask with three vertical necks. Center neck has 34/45 joint, and side necks have 24/40 joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.