Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, four neck, 3L, 24/40 center, (2)24/40, (1) 10/30 sides
SDP
Catalog No. 50973868
Encompass_Preferred
3L round bottom, heavy wall flask with four necks, arranged three in-line, and one 10/30 joint at 90. Center neck has a 24/40 joint, and vertical side necks have 24/40 joints.
Catalog No. 50-973-868 Supplier Ace Glass, Inc. Supplier No. 6953232
3L round bottom, heavy wall flask with four necks, arranged three in-line, and one 10/30 joint at 90. Center neck has a 24/40 joint, and vertical side necks have 24/40 joints.