Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, four neck, 3L, 24/40 center, (2)24/40, (1) #7 sides
SDP
Catalog No. 50973876
Encompass_Preferred
3L heavy wall, round bottom flask with four necks, arranged three in-line, and one #7 Ace-Thred neck. Center neck and angled side necks have 24/40 joints. Supplied with bushing and FETFE o-ring.
Catalog No. 50-973-876 Supplier Ace Glass, Inc. Supplier No. 6954285
3L heavy wall, round bottom flask with four necks, arranged three in-line, and one #7 Ace-Thred neck. Center neck and angled side necks have 24/40 joints. Supplied with bushing and FETFE o-ring.