Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, four neck, 3L, 24/40 center, 24/40 sides
SDP
Catalog No. 50973843
Encompass_Preferred
3L round bottom, heavy wall flask with four necks, arranged three in-line and one at 90. Center neck has a 24/40 joint, and angled side necks have 24/40 joints.
Catalog No. 50-973-843 Supplier Ace Glass, Inc. Supplier No. 6952232
3L round bottom, heavy wall flask with four necks, arranged three in-line and one at 90. Center neck has a 24/40 joint, and angled side necks have 24/40 joints.