Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, four neck, 5L, 34/45 center, 29/42 sides
SDP
Catalog No. 50973848
Encompass_Preferred
5L round bottom, heavy wall flask with four necks, arranged three in-line and one at 90. Center neck has a 34/45 joint, and angled side necks have 29/42 joints.
Catalog No. 50-973-848 Supplier Ace Glass, Inc. Supplier No. 6952257
5L round bottom, heavy wall flask with four necks, arranged three in-line and one at 90. Center neck has a 34/45 joint, and angled side necks have 29/42 joints.