Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, three neck, 5L, 35/25 center, 35/25 sides
SDP
Catalog No. 50973795
Encompass_Preferred
5L heavy wall, round bottom flask with three vertical necks: (1) 35/25 center neck, (2) 35/25 side necks.
Catalog No. 50-973-795 Supplier Ace Glass, Inc. Supplier No. 6950250
5L heavy wall, round bottom flask with three vertical necks: (1) 35/25 center neck, (2) 35/25 side necks.