Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, four neck, 5L, 45/50 center, 29/42 sides
SDP
Catalog No. 50973850
Encompass_Preferred
5L round bottom, heavy wall flask with four necks, arranged three in-line and one at 90. Center neck has a 45/50 joint, and angled side necks have 29/42 joints.
Catalog No. 50-973-850 Supplier Ace Glass, Inc. Supplier No. 6952263
5L round bottom, heavy wall flask with four necks, arranged three in-line and one at 90. Center neck has a 45/50 joint, and angled side necks have 29/42 joints.