Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, single neck, 5mL, 14/20
SDP
Catalog No. 50976817
Encompass_Preferred
5mL heavy walled, round bottom flask with a 14/20 single neck.
Catalog No. 50-976-817 Supplier Ace Glass, Inc. Supplier No. 9458202
5mL heavy walled, round bottom flask with a 14/20 single neck.