Learn More
Ace Glass, Inc. Flask, round bottom, 2000mL, 35/20, safety coated, replaces Buchi 40776, 25265
SDP
Catalog No. 50971546
Encompass_Preferred
2000mL round bottom replacement flask for Buchi rotary evaporators 40776 and 25265. Flask features a 35/20 single neck and is safety coated.
Catalog No. 50-971-546 Supplier Ace Glass, Inc. Supplier No. 399622
2000mL round bottom replacement flask for Buchi rotary evaporators 40776 and 25265. Flask features a 35/20 single neck and is safety coated.