Learn More
Ace Glass, Inc. Flask, round bottom, 500mL, 35/20, safety coated
SDP
Catalog No. 50971545
Encompass_Preferred
500mL round bottom replacement flask for Buchi rotary evaporators. Flask features a 35/20 single neck and is safety coated.
Catalog No. 50-971-545 Supplier Ace Glass, Inc. Supplier No. 399608
500mL round bottom replacement flask for Buchi rotary evaporators. Flask features a 35/20 single neck and is safety coated.