Learn More
Chemglass Life Sciences Flask, Shake, 1000mL, 4 Baffles, Constriction, Delong Neck, Approx OD x Height (mm): 130 x 230
SDP
Catalog No. 501532274
Encompass_Preferred
Flask, Shake, 1000mL, 4 Baffles, Constriction, Delong Neck, Approx OD x Height (mm): 130 x 230. Flask has 4 bottom baffles and a constriction just below the neck to prevent splashing from reaching the closure.
Catalog No. 50-153-2274 Supplier Chemglass Life Sciences Supplier No. CLS205006
Flask, Shake, 1000mL, 4 Baffles, Constriction, Delong Neck, Approx OD x Height (mm): 130 x 230. Flask has 4 bottom baffles and a constriction just below the neck to prevent splashing from reaching the closure.