Learn More
Chemglass Life Sciences Flask, Shake, 2000mL, Baffled, Delong Neck, Approx OD x Height (mm): 155 x 300
SDP
Catalog No. 501532267
Encompass_Preferred
Flask, Shake, 2000mL, Baffled, Delong Neck, Approx OD x Height (mm): 155 x 300. Baffle indents on the lower outside edge of the flask to help oxygen transfer to the culture medium when used on an orbital shaker. Delong style neck is made to fit Stainless Steel or plastic closures, while the longer neck reduces the amount of splashing that would reach the closure.
Catalog No. 50-153-2267 Supplier Chemglass Life Sciences Supplier No. CLS204408
Flask, Shake, 2000mL, Baffled, Delong Neck, Approx OD x Height (mm): 155 x 300. Baffle indents on the lower outside edge of the flask to help oxygen transfer to the culture medium when used on an orbital shaker. Delong style neck is made to fit Stainless Steel or plastic closures, while the longer neck reduces the amount of splashing that would reach the closure.