Learn More
Chemglass Life Sciences Flask, Shake, 250mL, 4 Baffles, Constriction, Delong Neck, Approx OD x Height (mm): 80 x 155
SDP
Catalog No. 501532272
Encompass_Preferred
Flask, Shake, 250mL, 4 Baffles, Constriction, Delong Neck, Approx OD x Height (mm): 80 x 155. Flask has 4 bottom baffles and a constriction just below the neck to prevent splashing from reaching the closure.
Catalog No. 50-153-2272 Supplier Chemglass Life Sciences Supplier No. CLS205002
Flask, Shake, 250mL, 4 Baffles, Constriction, Delong Neck, Approx OD x Height (mm): 80 x 155. Flask has 4 bottom baffles and a constriction just below the neck to prevent splashing from reaching the closure.