Learn More
Ace Glass, Inc. Flask, shake, 250mL, no baffles, 38mm OD long neck, cs/12
SDP
Catalog No. NC1718912
Encompass_Preferred
Flask, shake, 250mL, no baffles, 38mm OD long neck, cs/12
Catalog No. NC1718912 Supplier Ace Glass, Inc. Supplier No. 388419
Flask, shake, 250mL, no baffles, 38mm OD long neck, cs/12