Learn More
Chemglass Life Sciences 125mL Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 145 x 65
SDP
Catalog No. 50143857
Encompass_Preferred
125mL Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 145 x 65. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure.
Catalog No. 50-143-857 Supplier Chemglass Life Sciences Supplier No. CLS203603
125mL Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 145 x 65. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure.