Learn More
Chemglass Life Sciences 1L Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 130 x 230
SDP
Catalog No. 50143661
Encompass_Preferred
1L Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 130 x 230. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure.
Catalog No. 50-143-661 Supplier Chemglass Life Sciences Supplier No. CLS203607
1L Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 130 x 230. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure.