Learn More
Chemglass Life Sciences 25mL Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 40 x 90
SDP
Catalog No. 50143861
Encompass_Preferred
25mL Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 40 x 90. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure.
Catalog No. 50-143-861 Supplier Chemglass Life Sciences Supplier No. CLS203601
25mL Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 40 x 90. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure.