Learn More
Chemglass Life Sciences 2L Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 160 x 280
SDP
Catalog No. 50143674
Encompass_Preferred
2L Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 160 x 280. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure.
Catalog No. 50-143-674 Supplier Chemglass Life Sciences Supplier No. CLS203608
2L Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 160 x 280. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure.