Learn More
Chemglass Life Sciences 4L Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 205 x 370
SDP
Catalog No. 50143827
Encompass_Preferred
4L Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 205 x 370. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure.
Catalog No. 50-143-827 Supplier Chemglass Life Sciences Supplier No. CLS203610
4L Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 205 x 370. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure.