Learn More
Chemglass Life Sciences 50mL Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 50 x 110
SDP
Catalog No. 50143858
Encompass_Preferred
50mL Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 50 x 110. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure.
Catalog No. 50-143-858 Supplier Chemglass Life Sciences Supplier No. CLS203602
50mL Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 50 x 110. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure.