Learn More
Chemglass Life Sciences 6L Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 250 x 380
SDP
Catalog No. 50143710
Encompass_Preferred
6L Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 250 x 380. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure.
Catalog No. 50-143-710 Supplier Chemglass Life Sciences Supplier No. CLS203611
6L Shake Flask Only, No Baffles, Delong Neck, Approx OD x Height (mm): 250 x 380. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure.