Learn More
Chemglass Life Sciences 1L Shake Flask, No Baffles, Delong Neck, with Stainless Steel Closure, Approx OD x Height (mm): 130 x 230
SDP
Catalog No. 50143669
Encompass_Preferred
1L Shake Flask, No Baffles, Delong Neck, with Stainless Steel Closure, Approx OD x Height (mm): 130 x 230. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure. Supplied with a stainless steel closure.
Catalog No. 50-143-669 Supplier Chemglass Life Sciences Supplier No. CLS203807
1L Shake Flask, No Baffles, Delong Neck, with Stainless Steel Closure, Approx OD x Height (mm): 130 x 230. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure. Supplied with a stainless steel closure.