Learn More
Chemglass Life Sciences 6L Shake Flask, No Baffles, Delong Neck, with Stainless Steel Closure, Approx OD x Height (mm): 250 x 380
SDP
Catalog No. 50143687
Encompass_Preferred
6L Shake Flask, No Baffles, Delong Neck, with Stainless Steel Closure, Approx OD x Height (mm): 250 x 380. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure. Supplied with a stainless steel closure.
Catalog No. 50-143-687 Supplier Chemglass Life Sciences Supplier No. CLS203811
6L Shake Flask, No Baffles, Delong Neck, with Stainless Steel Closure, Approx OD x Height (mm): 250 x 380. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure. Supplied with a stainless steel closure.