Learn More
Ace Glass, Inc. Flask, shake, 125mL, three deep baffles, 25mm OD long neck, cs/12
SDP
Catalog No. 50971414
Encompass_Preferred
125mL cell culture shake flask with three outside edge baffles. The baffles are deeper that a standard flask to enhance mixing and oxygen transfer to media. The flask has a 25mm O.D. long neck to accomm ODate either plastic or stainless steel closures.
Catalog No. 50-971-414 Supplier Ace Glass, Inc. Supplier No. 388707
125mL cell culture shake flask with three outside edge baffles. The baffles are deeper that a standard flask to enhance mixing and oxygen transfer to media. The flask has a 25mm O.D. long neck to accomm ODate either plastic or stainless steel closures.