Learn More
Ace Glass, Inc. Flask, shake, 125mL, baffled, #5 stopper size beaded neck, cs/12
SDP
Catalog No. 50971420
Encompass_Preferred
125mL shake style culture flask with a #5 stopper size beaded neck, and three bottom edge baffles to enhance mixing and oxygen transfer to media.
Catalog No. 50-971-420 Supplier Ace Glass, Inc. Supplier No. 388914
125mL shake style culture flask with a #5 stopper size beaded neck, and three bottom edge baffles to enhance mixing and oxygen transfer to media.