Learn More
Ace Glass, Inc. Flask, shake, 125mL, no baffles, 25mm OD long neck
SDP
Catalog No. 50971407
Encompass_Preferred
125mL shake style culture flask without baffles. Flask has a 25mm O.D. long neck, and accomm ODates either plastic or stainless steel closures.
Catalog No. 50-971-407 Supplier Ace Glass, Inc. Supplier No. 388415
125mL shake style culture flask without baffles. Flask has a 25mm O.D. long neck, and accomm ODates either plastic or stainless steel closures.