Learn More
Ace Glass, Inc. Flask, shake, 125mL, three side baffles, 25mm OD Long Neck
SDP
Catalog No. 50971401
Encompass_Preferred
125mL shake flask with triple side baffles for cell culture applications. Flask has a 25mm O.D. long style straight neck to accomm ODate plastic and stainless steel closures. The flask is triple baffled on the bottom edge to increase mixing and oxygen transfer to the media.
Catalog No. 50-971-401 Supplier Ace Glass, Inc. Supplier No. 388305
125mL shake flask with triple side baffles for cell culture applications. Flask has a 25mm O.D. long style straight neck to accomm ODate plastic and stainless steel closures. The flask is triple baffled on the bottom edge to increase mixing and oxygen transfer to the media.