Learn More
Ace Glass, Inc. Flask, shake, 1L, baffled, #7 stopper size beaded neck
SDP
Catalog No. 50971422
Encompass_Preferred
1L shake style culture flask with a #7 stopper size beaded neck, and three bottom edge baffles to enhance mixing and oxygen transfer to media.
Catalog No. 50-971-422 Supplier Ace Glass, Inc. Supplier No. 388924
1L shake style culture flask with a #7 stopper size beaded neck, and three bottom edge baffles to enhance mixing and oxygen transfer to media.