Learn More
Ace Glass, Inc. Flask, shake, 250mL, baffled, #6 stopper size beaded neck, cs/12
SDP
Catalog No. 50978213
Encompass_Preferred
250mL shake style culture flask with a #6 stopper size beaded neck, and three bottom edge baffles to enhance mixing and oxygen transfer to media.
Catalog No. 50-978-213 Supplier Ace Glass, Inc. Supplier No. 388918
250mL shake style culture flask with a #6 stopper size beaded neck, and three bottom edge baffles to enhance mixing and oxygen transfer to media.