Learn More
Ace Glass, Inc. Flask, shake, 250mL, (3) side & (3) bottom deep baffles, 38mmlong neck
SDP
Catalog No. 50978215
Encompass_Preferred
250mL cell culture shake flask with three bottom edge baffles and three bottom baffles to greatly enhance mixing of cells in suspension and gas transfer to media. Ideal for use with reciprocating baths and shakers. Long style, straight neck accomodates either plastic or stainless steel closures.
Catalog No. 50-978-215 Supplier Ace Glass, Inc. Supplier No. 389106
250mL cell culture shake flask with three bottom edge baffles and three bottom baffles to greatly enhance mixing of cells in suspension and gas transfer to media. Ideal for use with reciprocating baths and shakers. Long style, straight neck accomodates either plastic or stainless steel closures.