Learn More
Ace Glass, Inc. Flask, shake, 25mL, no baffles, 18mm OD long neck, cs/24
SDP
Catalog No. 50978208
Encompass_Preferred
25mL shake style culture flask without baffles. Flask has an 18mm O.D. long neck, and accomm ODates either plastic or stainless steel closures.
Catalog No. 50-978-208 Supplier Ace Glass, Inc. Supplier No. 388405
25mL shake style culture flask without baffles. Flask has an 18mm O.D. long neck, and accomm ODates either plastic or stainless steel closures.