Learn More
Ace Glass, Inc. Flask, shake, 2L, three deep baffles, 38mm OD long neck, cs/3
SDP
Catalog No. 50978212
Encompass_Preferred
2L cell culture shake flask with three outside edge baffles. The baffles are deeper that a standard flask to enhance mixing and oxygen transfer to media. The flask has a 38mm O.D. long neck to accomm ODate either plastic or stainless steel closures.
Catalog No. 50-978-212 Supplier Ace Glass, Inc. Supplier No. 388719
2L cell culture shake flask with three outside edge baffles. The baffles are deeper that a standard flask to enhance mixing and oxygen transfer to media. The flask has a 38mm O.D. long neck to accomm ODate either plastic or stainless steel closures.