Learn More
Ace Glass, Inc. Flask, shake, 2L, no baffles, 38mm OD long neck, cs/3
SDP
Catalog No. 50971411
Encompass_Preferred
2L shake style culture flask without baffles. Flask has a 38mm O.D. long neck, and accomm ODates either plastic or stainless steel closures.
Catalog No. 50-971-411 Supplier Ace Glass, Inc. Supplier No. 388434
2L shake style culture flask without baffles. Flask has a 38mm O.D. long neck, and accomm ODates either plastic or stainless steel closures.