Learn More
Ace Glass, Inc. Flask, shake, 2L, three side baffles, 38mm OD long neck, cs/3
SDP
Catalog No. 50971405
Encompass_Preferred
2L shake flask with triple side baffles for cell culture applications. Flask has a 38mm O.D. long style straight neck to accomm ODate plastic and stainless steel closures. The flask is triple baffled on the bottom edge to increase mixing and oxygen transfer to the media.
Catalog No. 50-971-405 Supplier Ace Glass, Inc. Supplier No. 388315
2L shake flask with triple side baffles for cell culture applications. Flask has a 38mm O.D. long style straight neck to accomm ODate plastic and stainless steel closures. The flask is triple baffled on the bottom edge to increase mixing and oxygen transfer to the media.