Learn More
Ace Glass, Inc. Flask, shake, 300mL, no baffles, 38mm OD long neck, cs/12
SDP
Catalog No. 50978209
Encompass_Preferred
300mL shake style culture flask without baffles. Flask has a 38mm O.D. long neck, and accomm ODates either plastic or stainless steel closures.
Catalog No. 50-978-209 Supplier Ace Glass, Inc. Supplier No. 388423
300mL shake style culture flask without baffles. Flask has a 38mm O.D. long neck, and accomm ODates either plastic or stainless steel closures.