Learn More
Ace Glass, Inc. Flask, shake, 3L, (3) side & (3) bottom deep baffles, 38mm longneck, cs/3
SDP
Catalog No. 50978216
Encompass_Preferred
3L cell culture shake flask with three bottom edge baffles and three bottom baffles to greatly enhance mixing of cells in suspension and gas transfer to media. Ideal for use with reciprocating baths and shakers. Long style, straight neck accomodates either plastic or stainless steel closures.
Catalog No. 50-978-216 Supplier Ace Glass, Inc. Supplier No. 389125
3L cell culture shake flask with three bottom edge baffles and three bottom baffles to greatly enhance mixing of cells in suspension and gas transfer to media. Ideal for use with reciprocating baths and shakers. Long style, straight neck accomodates either plastic or stainless steel closures.