Learn More
Ace Glass, Inc. Flask, shake, 4L, no baffles, 38mm OD long neck, cs/3
SDP
Catalog No. 50978210
Encompass_Preferred
4L shake style culture flask without baffles. Flask has a 38mm O.D. long neck, and accomm ODates either plastic or stainless steel closures.
Catalog No. 50-978-210 Supplier Ace Glass, Inc. Supplier No. 388437
4L shake style culture flask without baffles. Flask has a 38mm O.D. long neck, and accomm ODates either plastic or stainless steel closures.