Learn More
Ace Glass, Inc. Flask, shake, 4L, (3) side & (3) bottom deep baffles, 38mm longneck, cs/3
SDP
Catalog No. 50971430
Encompass_Preferred
4L cell culture shake flask with three bottom edge baffles and three bottom baffles to greatly enhance mixing of cells in suspension and gas transfer to media. Ideal for use with reciprocating baths and shakers. Long style, straight neck accomodates either plastic or stainless steel closures.
Catalog No. 50-971-430 Supplier Ace Glass, Inc. Supplier No. 389129
4L cell culture shake flask with three bottom edge baffles and three bottom baffles to greatly enhance mixing of cells in suspension and gas transfer to media. Ideal for use with reciprocating baths and shakers. Long style, straight neck accomodates either plastic or stainless steel closures.