Learn More
Ace Glass, Inc. Flask, shake, 500mL, three deep baffles, 38mm OD long neck, cs/12
SDP
Catalog No. 50971416
Encompass_Preferred
500mL cell culture shake flask with three outside edge baffles. The baffles are deeper that a standard flask to enhance mixing and oxygen transfer to media. The flask has a 38mm O.D. long neck to accomm ODate either plastic or stainless steel closures.
Catalog No. 50-971-416 Supplier Ace Glass, Inc. Supplier No. 388713
500mL cell culture shake flask with three outside edge baffles. The baffles are deeper that a standard flask to enhance mixing and oxygen transfer to media. The flask has a 38mm O.D. long neck to accomm ODate either plastic or stainless steel closures.