Learn More
Ace Glass, Inc. Flask, shake, 500mL, baffled, #6 stopper size beaded neck, cs/12
SDP
Catalog No. 50971421
Encompass_Preferred
500mL shake style culture flask with a #6 stopper size beaded neck, and three bottom edge baffles to enhance mixing and oxygen transfer to media.
Catalog No. 50-971-421 Supplier Ace Glass, Inc. Supplier No. 388922
500mL shake style culture flask with a #6 stopper size beaded neck, and three bottom edge baffles to enhance mixing and oxygen transfer to media.