Learn More
Ace Glass, Inc. Flask, shake, 500mL, (3) side & (3) bottom deep baffles, 38mmlong neck, cs/12
SDP
Catalog No. 50971427
Encompass_Preferred
500mL cell culture shake flask with three bottom edge baffles and three bottom baffles to greatly enhance mixing of cells in suspension and gas transfer to media. Ideal for use with reciprocating baths and shakers. Long style, straight neck accomodates either plastic or stainless steel closures.
Catalog No. 50-971-427 Supplier Ace Glass, Inc. Supplier No. 389111
500mL cell culture shake flask with three bottom edge baffles and three bottom baffles to greatly enhance mixing of cells in suspension and gas transfer to media. Ideal for use with reciprocating baths and shakers. Long style, straight neck accomodates either plastic or stainless steel closures.